Ethyl 4-isopropoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

59119-500MG 59119-1G 59119-5G 59119-25G
77330-454EA 152.44 USD
77330-454 77345-458 77366-680 77388-380
Ethyl 4-isopropoxy-3-methylbenzoate ≥97%
Ethyl 4-isopropoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759002
CAS Number: 1350838-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR