4-Ethoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

59115-500MG 59115-1G 59115-5G 59115-25G
77330-450EA 138.02 USD
77330-450 77344-602 77366-676 77388-376
4-Ethoxy-3-methylbenzoic acid ≥97%
4-Ethoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13920122
CAS Number: 92315-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR