(4-Ethoxy-3-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

59111-500MG 59111-1G 59111-5G 59111-25G
77330-446EA 178.19 USD
77330-446 77346-664 77366-672 77388-372
(4-Ethoxy-3-methylphenyl)(methyl)sulfane ≥95%
(4-Ethoxy-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD18375530
CAS Number: 1378808-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR