(4-Methoxy-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59110-1G 59110-5G 59110-25G
77341-556EA 92.7 USD
77341-556 77366-670 77378-720
(4-Methoxy-3-methylphenyl)(methyl)sulfane ≥97%
(4-Methoxy-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₂OS
MW: 168.256 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010335
CAS Number: 50390-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR