1-(Benzyloxy)-4-iodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59107-500MG 59107-1G 59107-5G 59107-25G
77330-442EA 257.5 USD
77330-442 77354-622 77366-668 77388-370
1-(Benzyloxy)-4-iodo-2-methylbenzene ≥97%
1-(Benzyloxy)-4-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃IO
Storage Temperature: Ambient
MDL Number: MFCD32649106
CAS Number: 516521-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR