1-Chloro-2-fluoro-3,4-diiodobenzene ≥97%

Supplier: AOBChem USA

Warning

59101-500MG 59101-1G 59101-5G 59101-25G
77330-436EA 90.64 USD
77330-436 77342-398 77366-662 77378-716
1-Chloro-2-fluoro-3,4-diiodobenzene ≥97%
1-Chloro-2-fluoro-3,4-diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClFI₂
MW: 382.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751404
CAS Number: 1805029-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR