2-Bromo-4-chloro-3-fluoro-1-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

59100-500MG 59100-1G 59100-5G 59100-25G
77330-434EA 169.95 USD
77330-434 77346-352 77366-660 77388-364
2-Bromo-4-chloro-3-fluoro-1-iodobenzene ≥95%
2-Bromo-4-chloro-3-fluoro-1-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
Storage Temperature: Ambient
MDL Number: MFCD34758998
CAS Number: 2385938-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR