1-Bromo-2-chloro-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

59090-500MG 59090-1G 59090-5G 59090-25G
77330-432EA 142.14 USD
77330-432 77344-976 77366-658 77388-362
1-Bromo-2-chloro-3-(methylsulfonyl)benzene ≥97%
1-Bromo-2-chloro-3-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₆BrClO₂S
MW: 269.543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758989
CAS Number: 871352-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR