Methyl 2-chloro-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

59088-500MG 59088-1G 59088-5G
77339-222EA 568.56 USD
77339-222 77354-618 77378-714
Methyl 2-chloro-3-(methylthio)benzoate ≥97%
Methyl 2-chloro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂S
MW: 216.685 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489085
CAS Number: 1823024-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR