4-Bromo-3-chloro-2,4'-difluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

59085-500MG 59085-1G 59085-5G 59085-25G
77330-430EA 204.97 USD
77330-430 77354-616 77366-656 77388-360
4-Bromo-3-chloro-2,4'-difluoro-1,1'-biphenyl ≥97%
4-Bromo-3-chloro-2,4'-difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD34758987
CAS Number: 2755718-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR