2,4-Dichloro-N-cyclopropyl-3-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

59072-500MG 59072-1G 59072-5G
77338-774EA 517.06 USD
77338-774 77354-612 77378-710
2,4-Dichloro-N-cyclopropyl-3-fluorobenzamide ≥97%
2,4-Dichloro-N-cyclopropyl-3-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂FNO
MW: 248.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758978
CAS Number: 2755723-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR