2,4-Dichloro-3-fluorobenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
59068-500MG
59068-1G
59068-5G
77338-534EA
496.46
USD
77338-534
77354-608
77378-706
2,4-Dichloro-3-fluorobenzamide ≥97%
2,4-Dichloro-3-fluorobenzamide
Precaución: for Research Use Only
Formula:
C₇H₄Cl₂FNO Storage Temperature: Ambient |
MDL Number:
MFCD28785957 CAS Number: 1803718-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...