1-Bromo-2-chloro-4-iodo-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

59064-1G 59064-5G 59064-25G
77341-828EA 105.06 USD
77341-828 77366-652 77378-704
1-Bromo-2-chloro-4-iodo-3-methoxybenzene ≥97%
1-Bromo-2-chloro-4-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClIO
MW: 347.375 g/mol
Storage Temperature: Ambient
CAS Number: 2385813-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR