1,4-Dibromo-2-chloro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

59063-1G 59063-5G 59063-25G
77341-194EA 74.16 USD
77341-194 77366-650 77378-702
1,4-Dibromo-2-chloro-3-methoxybenzene ≥97%
1,4-Dibromo-2-chloro-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂ClO
MW: 300.375 g/mol
Storage Temperature: Ambient
CAS Number: 174913-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR