1-Bromo-2-chloro-3,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

59062-500MG 59062-1G 59062-5G
77338-524EA 494.4 USD
77338-524 77354-606 77378-700
1-Bromo-2-chloro-3,4-dimethoxybenzene ≥97%
1-Bromo-2-chloro-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO₂
MW: 251.505 g/mol
Storage Temperature: Ambient
CAS Number: 343867-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR