(2-Chloro-3-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59060-500MG 59060-1G 59060-5G
77339-096EA 550.02 USD
77339-096 77354-604 77378-698
(2-Chloro-3-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%
(2-Chloro-3-fluoro-4-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
Storage Temperature: Ambient
MDL Number: MFCD28503563
CAS Number: 1935403-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR