3-Fluoro-N-methoxy-5-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

59049-500MG 59049-1G 59049-5G 59049-25G
77330-422EA 179.22 USD
77330-422 77346-730 77366-648 77388-356
3-Fluoro-N-methoxy-5-methylbenzamide ≥97%
3-Fluoro-N-methoxy-5-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD26798242
CAS Number: 1566276-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR