2-Isopropoxy-3,4-dimethoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

59038-500MG 59038-1G 59038-5G
77336-428EA 387.28 USD
77336-428 77354-596 77378-690
2-Isopropoxy-3,4-dimethoxybenzoic acid ≥95%
2-Isopropoxy-3,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₅
Storage Temperature: Ambient
MDL Number: MFCD34758970
CAS Number: 5651-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR