(3-(Benzyloxy)-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59034-500MG 59034-1G 59034-5G
77338-268EA 470.71 USD
77338-268 77354-590 77378-684
(3-(Benzyloxy)-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(3-(Benzyloxy)-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₄OS
MW: 316.314 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758967
CAS Number: 2755722-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR