3-(Benzyloxy)-2-fluoro-6-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

59030-500MG 59030-1G 59030-5G
77338-772EA 517.06 USD
77338-772 77354-588 77378-682
3-(Benzyloxy)-2-fluoro-6-(trifluoromethyl)benzoic acid ≥97%
3-(Benzyloxy)-2-fluoro-6-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₀F₄O₃
MW: 314.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758963
CAS Number: 2755719-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR