2-Fluoro-3-iodo-1-isopropoxy-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

59027-500MG 59027-1G 59027-5G
77339-132EA 553.11 USD
77339-132 77354-584 77378-678
2-Fluoro-3-iodo-1-isopropoxy-4-(trifluoromethyl)benzene ≥95%
2-Fluoro-3-iodo-1-isopropoxy-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉F₄IO
MW: 348.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758960
CAS Number: 2807447-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR