(2-Fluoro-3-isopropoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59026-500MG 59026-1G 59026-5G
77339-442EA 590.19 USD
77339-442 77354-582 77378-676
(2-Fluoro-3-isopropoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-isopropoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₄OS
MW: 268.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758959
CAS Number: 2807441-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR