(2-Fluoro-3-isopropoxy-6-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59025-500MG 59025-1G 59025-5G
77339-762EA 631.39 USD
77339-762 77354-580 77378-674
(2-Fluoro-3-isopropoxy-6-(trifluoromethyl)phenyl)methanol ≥97%
(2-Fluoro-3-isopropoxy-6-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34758958
CAS Number: 2807455-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR