2-Fluoro-3-isopropoxy-6-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

59024-500MG 59024-1G 59024-5G
77339-786EA 638.6 USD
77339-786 77354-578 77378-672
2-Fluoro-3-isopropoxy-6-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-3-isopropoxy-6-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₄O₂
MW: 250.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758957
CAS Number: 2755718-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR