Methyl 2-fluoro-3-isopropoxy-6-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

59023-500MG 59023-1G 59023-5G
77339-238EA 569.59 USD
77339-238 77354-576 77378-670
Methyl 2-fluoro-3-isopropoxy-6-(trifluoromethyl)benzoate ≥95%
Methyl 2-fluoro-3-isopropoxy-6-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₄O₃
MW: 280.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758956
CAS Number: 2901065-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR