1-Ethoxy-2-fluoro-3-iodo-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

59019-500MG 59019-1G 59019-5G
77339-826EA 653.02 USD
77339-826 77354-572 77378-666
1-Ethoxy-2-fluoro-3-iodo-4-(trifluoromethyl)benzene ≥97%
1-Ethoxy-2-fluoro-3-iodo-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇F₄IO
MW: 334.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758952
CAS Number: 2384696-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR