(3-Ethoxy-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
59018-500MG
59018-1G
59018-5G
77338-608EA
503.67
USD
77338-608
77354-570
77378-664
(3-Ethoxy-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(3-Ethoxy-2-fluoro-6-(trifluoromethyl)phenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₀H₁₀F₄OS Storage Temperature: Ambient |
MDL Number:
MFCD34758951 CAS Number: 2710879-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...