2-Chloro-6-fluoro-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58993-500MG 58993-1G 58993-5G 58993-25G
77330-420EA 229.69 USD
77330-420 77354-560 77366-646 77388-354
2-Chloro-6-fluoro-3-(methylthio)benzaldehyde ≥97%
2-Chloro-6-fluoro-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFOS
MW: 204.649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758941
CAS Number: 2807471-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR