(2-(Benzyloxy)-4-chlorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

58976-500MG 58976-1G 58976-5G 58976-25G
77330-416EA 147.29 USD
77330-416 77345-218 77366-642 77388-350
(2-(Benzyloxy)-4-chlorophenyl)(methyl)sulfane ≥97%
(2-(Benzyloxy)-4-chlorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClOS
Storage Temperature: Ambient
MDL Number: MFCD34758936
CAS Number: 2764728-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR