4-Isopropoxy-3-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

58971-500MG 58971-1G 58971-5G
77335-768EA 365.65 USD
77335-768 77354-554 77378-652
4-Isopropoxy-3-(trifluoromethoxy)benzoic acid ≥97%
4-Isopropoxy-3-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₄
MW: 264.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758933
CAS Number: 2807456-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR