4-Isopropoxy-3-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58969-500MG 58969-1G 58969-25G 58969-5G
77330-414EA 285.31 USD
77330-414 77354-552 77388-348 77378-650
4-Isopropoxy-3-(trifluoromethoxy)benzaldehyde ≥97%
4-Isopropoxy-3-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₃
MW: 248.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29111331
CAS Number: 1193467-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR