2-(Benzyloxy)-3-chloro-1,4-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

58960-1G 58960-5G 58960-25G
77341-430EA 85.49 USD
77341-430 77366-640 77378-648
2-(Benzyloxy)-3-chloro-1,4-difluorobenzene ≥95%
2-(Benzyloxy)-3-chloro-1,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉ClF₂O
MW: 254.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564587
CAS Number: 2734772-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR