2-Ethoxy-3-fluoro-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

58953-500MG 58953-1G 58953-5G
77339-192EA 562.38 USD
77339-192 77354-550 77378-646
2-Ethoxy-3-fluoro-5-(trifluoromethyl)benzoic acid ≥97%
2-Ethoxy-3-fluoro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758925
CAS Number: 2701612-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR