2-(Benzyloxy)-1-fluoro-3-iodo-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

58917-500MG 58917-1G 58917-5G
77339-094EA 550.02 USD
77339-094 77354-544 77378-642
2-(Benzyloxy)-1-fluoro-3-iodo-5-(trifluoromethyl)benzene ≥97%
2-(Benzyloxy)-1-fluoro-3-iodo-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉F₄IO
MW: 396.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758916
CAS Number: 2764728-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR