1-Fluoro-3-iodo-2-isopropoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

58916-500MG 58916-1G 58916-5G
77339-638EA 605.64 USD
77339-638 77354-542 77378-640
1-Fluoro-3-iodo-2-isopropoxy-5-(trifluoromethyl)benzene ≥97%
1-Fluoro-3-iodo-2-isopropoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉F₄IO
Storage Temperature: Ambient
MDL Number: MFCD34758915
CAS Number: 2764730-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR