2-Ethoxy-1-fluoro-3-iodo-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

58915-500MG 58915-1G 58915-5G
77338-384EA 484.1 USD
77338-384 77354-540 77378-638
2-Ethoxy-1-fluoro-3-iodo-5-(trifluoromethyl)benzene ≥97%
2-Ethoxy-1-fluoro-3-iodo-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇F₄IO
Storage Temperature: Ambient
MDL Number: MFCD34758914
CAS Number: 2384300-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR