2',3',4'-Trimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58911-500MG 58911-1G 58911-5G
77337-298EA 410.97 USD
77337-298 77354-538 77378-636
2',3',4'-Trimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2',3',4'-Trimethoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₄
MW: 272.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11932522
CAS Number: 1181591-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR