Methyl 2-ethoxy-3,4-dimethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

58903-500MG 58903-1G 58903-5G
77338-266EA 470.71 USD
77338-266 77354-534 77378-632
Methyl 2-ethoxy-3,4-dimethoxybenzoate ≥97%
Methyl 2-ethoxy-3,4-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₅
Storage Temperature: Ambient
MDL Number: MFCD28059985
CAS Number: 2117093-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR