1-Bromo-2-ethoxy-3,4-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

58899-500MG 58899-1G 58899-5G
77336-914EA 399.64 USD
77336-914 77354-532 77378-630
1-Bromo-2-ethoxy-3,4-dimethoxybenzene ≥95%
1-Bromo-2-ethoxy-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₃
Storage Temperature: Ambient
MDL Number: MFCD34758906
CAS Number: 2385860-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR