5-(4-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58876-500MG 58876-1G 58876-5G
77330-410EA 340.93 USD
77330-410 77354-530 77378-628
5-(4-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(4-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆ClF₃N₂S
MW: 278.681 g/mol
Storage Temperature: Ambient
CAS Number: 2901083-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR