1-Bromo-2-isopropoxy-3,4-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

58874-500MG 58874-1G 58874-25G 58874-5G
77330-408EA 296.64 USD
77330-408 77354-528 77388-344 77378-626
1-Bromo-2-isopropoxy-3,4-dimethoxybenzene ≥95%
1-Bromo-2-isopropoxy-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD27941743
CAS Number: 114605-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR