(3-Fluoro-5-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
58866-500MG
58866-1G
58866-5G
58866-25G
77330-406EA
95.79
USD
77330-406
77342-570
77366-634
77378-620
(3-Fluoro-5-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(3-Fluoro-5-methylphenyl)(4-methylpiperazin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₃H₁₇FN₂O MW: 236.285 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26984970 CAS Number: 1779329-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...