N-Cyclohexyl-3-fluoro-5-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

58861-500MG 58861-1G 58861-5G 58861-25G
77330-404EA 85.49 USD
77330-404 77342-226 77366-632 77378-618
N-Cyclohexyl-3-fluoro-5-methylbenzamide ≥97%
N-Cyclohexyl-3-fluoro-5-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO
Storage Temperature: Ambient
MDL Number: MFCD26984971
CAS Number: 1710413-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR