N-Cyclopropyl-3-fluoro-5-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

58860-500MG 58860-1G 58860-5G 58860-25G
77330-402EA 136.99 USD
77330-402 77344-542 77366-630 77388-342
N-Cyclopropyl-3-fluoro-5-methylbenzamide ≥97%
N-Cyclopropyl-3-fluoro-5-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD26798234
CAS Number: 1564811-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR