2-Bromo-6-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58846-500MG 58846-1G 58846-5G 58846-25G
77330-398EA 187.46 USD
77330-398 77354-522 77366-626 77388-338
2-Bromo-6-methoxy-4'-methyl-1,1'-biphenyl ≥97%
2-Bromo-6-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758878
CAS Number: 2764731-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR