2'-Bromo-6'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58845-500MG 58845-1G 58845-5G 58845-25G
77330-396EA 225.57 USD
77330-396 77354-520 77366-624 77388-336
2'-Bromo-6'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Bromo-6'-methoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO₂
MW: 291.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758877
CAS Number: 2755723-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR