Ethyl 3-ethoxy-2-fluoro-5-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

58844-500MG 58844-1G 58844-5G
77330-394EA 345.05 USD
77330-394 77354-518 77378-612
Ethyl 3-ethoxy-2-fluoro-5-methoxybenzoate ≥95%
Ethyl 3-ethoxy-2-fluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₄
MW: 242.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758876
CAS Number: 2704573-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR