(3-Ethoxy-2-fluoro-5-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

58835-500MG 58835-1G 58835-5G 58835-25G
77330-384EA 258.53 USD
77330-384 77354-504 77366-618 77388-328
(3-Ethoxy-2-fluoro-5-methoxyphenyl)(methyl)sulfane ≥97%
(3-Ethoxy-2-fluoro-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂S
MW: 216.272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758867
CAS Number: 2705651-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR