2-Chloro-3-fluoro-4-(p-tolyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

58824-500MG 58824-1G 58824-5G 58824-25G
77330-382EA 233.81 USD
77330-382 77354-502 77366-616 77388-326
2-Chloro-3-fluoro-4-(p-tolyl)pyridine ≥97%
2-Chloro-3-fluoro-4-(p-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₉ClFN
MW: 221.658 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758861
CAS Number: 2764730-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR