3'-Chloro-5'-fluoro-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58820-500MG 58820-1G 58820-5G 58820-25G
77330-378EA 178.19 USD
77330-378 77346-662 77366-612 77388-322
3'-Chloro-5'-fluoro-2-methyl-1,1'-biphenyl ≥97%
3'-Chloro-5'-fluoro-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClF
MW: 220.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758858
CAS Number: 2807450-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR